Assay Details
| Gene Symbol: Zdhhc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023740.2 | NP_076229.2 | 8 - 9 | 1117 | 122 | |
| GenBank mRNA | AF176814.1 | - | 8 - 9 | 1090 | 122 | |
| AK005255.1 | - | 7 - 8 | 849 | 122 | ||
| AK151505.1 | - | 8 - 9 | 1099 | 122 | ||
| AK168042.1 | - | 8 - 9 | 1092 | 122 | ||
| AK181099.1 | - | 1 - 2 | 81 | 122 | ||
| AK193583.1 | - | 1 - 2 | 81 | 122 | ||
| AK206660.1 | - | 1 - 2 | 81 | 122 | ||
| AK219891.1 | - | 1 - 2 | 81 | 122 | ||
| BC010835.1 | - | 8 - 9 | 1081 | 4239913 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc16
Genomic Map