Assay Details
| Gene Symbol: Dtx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256096.1 | NP_001243025.1 | 4 - 5 | 1305 | 154 | |
| NM_001256097.1 | NP_001243026.1 | 5 - 6 | 1383 | 154 | ||
| NM_001256098.1 | NP_001243027.1 | 4 - 5 | 1305 | 154 | ||
| NM_023742.2 | NP_076231.1 | 4 - 5 | 1305 | 154 | ||
| GenBank mRNA | AB015423.1 | - | 4 - 5 | 1228 | 154 | |
| AB015424.1 | - | 4 - 5 | 1228 | 154 | ||
| AK012137.1 | - | 4 - 5 | 1301 | 154 | ||
| AK044105.1 | - | 5 - 6 | 1340 | 154 | ||
| AK054443.1 | - | 4 - 5 | 1295 | 154 | ||
| BC024925.1 | - | 4 - 5 | 1247 | 3711019 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dtx2
Genomic Map