Assay Details
| Gene Symbol: Ankrd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023816.2 | NP_076305.2 | 2 - 3 | 558 | 87 | |
| GenBank mRNA | AF294328.1 | - | 2 - 3 | 560 | 87 | |
| AK005925.1 | - | 2 - 3 | 532 | 87 | ||
| AK132751.1 | - | 2 - 3 | 559 | 87 | ||
| BC120815.1 | - | 2 - 3 | 336 | 87 | ||
| BC120817.1 | - | 2 - 3 | 333 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd36
Genomic Map