Assay Details
| Gene Symbol: Ankrd36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023816.2 | NP_076305.2 | 3 - 4 | 734 | 95 | |
| GenBank mRNA | AF294328.1 | - | 3 - 4 | 736 | 95 | |
| AK005925.1 | - | 3 - 4 | 708 | 95 | ||
| AK132751.1 | - | 3 - 4 | 735 | 95 | ||
| BC120815.1 | - | 3 - 4 | 512 | 95 | ||
| BC120817.1 | - | 3 - 4 | 509 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ankrd36
Genomic Map