Assay Details
| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023831.3 | NP_076320.2 | 2 - 3 | 261 | 66 | |
| GenBank mRNA | AJ249981.1 | - | 2 - 3 | 305 | 66 | |
| AK005360.1 | - | 2 - 3 | 269 | 66 | ||
| AK032880.1 | - | 2 - 3 | 262 | 66 | ||
| BC080764.1 | - | 2 - 3 | 203 | 6488976 | 66 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ249981.1,AK005360.1,AK032880.1,BC080764.1,NM_023831.3 transcripts.
Set Membership: |
|

Ift46
Genomic Map