Assay Details
| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023831.3 | NP_076320.2 | 10 - 11 | 1024 | 81 | |
| GenBank mRNA | AJ249981.1 | - | 10 - 11 | 1065 | 81 | |
| AK005360.1 | - | 10 - 11 | 1032 | 81 | ||
| AK032880.1 | - | 10 - 11 | 1025 | 81 | ||
| BC010326.1 | - | 9 - 10 | 823 | 3599905 | 81 | |
| BC080764.1 | - | 10 - 11 | 966 | 6488976 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift46
Genomic Map