Assay Details
| Gene Symbol: Tsc22d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023910.6 | NP_076399.4 | 2 - 3 | 1457 | 97 | |
| NM_029805.1 | NP_084081.1 | 2 - 3 | 1457 | 97 | ||
| GenBank mRNA | AF201286.1 | - | 2 - 3 | 1170 | 97 | |
| AK018735.2 | - | 2 - 3 | 1430 | 97 | ||
| AK040534.1 | - | 2 - 3 | 1394 | 97 | ||
| AK079510.1 | - | 2 - 3 | 1183 | 97 | ||
| AK080828.1 | - | 2 - 3 | 1472 | 97 | ||
| AK143608.1 | - | 2 - 3 | 1234 | 97 | ||
| AK144806.1 | - | 1 - 2 | 894 | 97 | ||
| AK150043.1 | - | 2 - 3 | 1426 | 97 | ||
| AK154153.1 | - | 2 - 3 | 1407 | 97 | ||
| AK154490.1 | - | 2 - 3 | 1057 | 97 | ||
| AK155712.1 | - | 2 - 3 | 1459 | 97 | ||
| AK170487.1 | - | 2 - 3 | 1446 | 97 | ||
| BC018544.1 | - | 1 - 2 | 929 | 3666221 | 97 | |
| BC023761.1 | - | 2 - 3 | 1361 | 5343927 | 97 | |
| BC145721.1 | - | 2 - 3 | 1420 | 40131024 | 97 | |
| BC145723.1 | - | 2 - 3 | 1420 | 40131026 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tsc22d4
Genomic Map