Assay Details
| Gene Symbol: Hsdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301367.1 | NP_001288296.1 | 6 - 7 | 790 | 79 | |
| NM_024255.3 | NP_077217.2 | 5 - 6 | 692 | 79 | ||
| GenBank mRNA | AK011910.1 | - | 5 - 6 | 650 | 79 | |
| AK085899.1 | - | 5 - 6 | 661 | 79 | ||
| AK136773.1 | - | 5 - 6 | 659 | 79 | ||
| AK144948.1 | - | 5 - 6 | 608 | 79 | ||
| AK145230.1 | - | 5 - 6 | 552 | 79 | ||
| AY725196.1 | - | 5 - 6 | 549 | 79 | ||
| BC002211.1 | - | 5 - 6 | 565 | 3490303 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsdl2
Genomic Map