Assay Details
| Gene Symbol: Mxra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024263.4 | NP_077225.4 | 6 - 7 | 1268 | 93 | |
| GenBank mRNA | AB040488.1 | - | 6 - 7 | 1188 | 93 | |
| AB052097.1 | - | 6 - 7 | 1146 | 93 | ||
| AK004732.1 | - | 6 - 7 | 1151 | 93 | ||
| BC002140.1 | - | 1 - 2 | 265 | 93 | ||
| BC026438.1 | - | 6 - 7 | 1190 | 4239626 | 93 | |
| EU849596.1 | - | 6 - 7 | 1106 | 93 | ||
| EU849597.1 | - | 6 - 7 | 1097 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mxra8
Genomic Map