Assay Details
| Gene Symbol: Mxra8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024263.4 | NP_077225.4 | 7 - 8 | 1301 | 68 | |
| GenBank mRNA | AB040488.1 | - | 7 - 8 | 1221 | 68 | |
| AB052097.1 | - | 7 - 8 | 1179 | 68 | ||
| AK004732.1 | - | 7 - 8 | 1184 | 68 | ||
| BC002140.1 | - | 2 - 3 | 298 | 68 | ||
| BC026438.1 | - | 7 - 8 | 1223 | 4239626 | 68 | |
| EU849596.1 | - | 7 - 8 | 1139 | 68 | ||
| EU849597.1 | - | 7 - 8 | 1130 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mxra8
Genomic Map