Assay Details
| Gene Symbol: Duoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025777.2 | NP_080053.1 | 1 - 2 | 294 | 69 | |
| GenBank mRNA | AK008816.1 | - | 1 - 2 | 294 | 69 | |
| AK018569.1 | - | 1 - 2 | 296 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585601 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585602 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107374 | BC031111.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107374 | NM_025777.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2107374 | AK018569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006644 | NM_025777.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006644 | AK018569.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393756 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 60781 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147944 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006644 | NM_025777.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006644 | AK018569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107374 | BC031111.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107374 | NM_025777.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2107374 | AK018569.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585578 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585571 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585573 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585580 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585577 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21585543 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147944 | - | - |
| Whole Mouse Genome G4122A | A_51_P365744 | BC031111.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P365744 | NM_025777.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P365744 | AK008816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P365744 | AK018569.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119694 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147944 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01294105 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585577 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585602 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585605 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585601 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585578 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585580 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585581 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21585603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21585588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01294090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01294099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585612 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21585606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01871532 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10475454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475449 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475450 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475451 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475452 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422137_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10475448 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486988 | - | - |
| Mouse Genome 430 2.0 Array | 1422137_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422137_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66811_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66811_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200014316.mm.1 | NM_025777.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014316.mm.1 | NM_025777.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014316.mm.1 | TC0200001846.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200014315.mm.1 | NM_025777.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014315.mm.1 | NM_025777.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200014315.mm.1 | TC0200001846.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422137_at | - | - |
| Mouse Expression Set 430 | 1422137_PM_at | - | - |
| Murine Genome U74v2 Set | 167708_at | - | - |
| Murine Genome U74v2 Set | 167708_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422137_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422137_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422137_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422137_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 720766 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7040048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007040048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2589859 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2589859 | - | - |
| mouse-6 v1.1 (Illumina) | scl20366.6.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7040048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20366.6.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7040048 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2589859 | - | - |

Duoxa2
Genomic Map