Assay Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026156.2 | NP_080432.1 | 3 - 4 | 342 | 69 | |
| GenBank mRNA | AK002890.1 | - | 3 - 4 | 343 | 69 | |
| AK031533.1 | - | 352 | 69 | |||
| AK087952.1 | - | 3 - 4 | 349 | 69 | ||
| AK132434.1 | - | 3 - 4 | 356 | 69 | ||
| AK145925.1 | - | 3 - 4 | 344 | 69 | ||
| AK199409.1 | - | 1 - 2 | 90 | 69 | ||
| AK214621.1 | - | 1 - 2 | 90 | 69 | ||
| BC021341.1 | - | 3 - 4 | 340 | 5066146 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xab2
Genomic Map