Assay Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026156.2 | NP_080432.1 | 5 - 6 | 680 | 71 | |
| GenBank mRNA | AK002890.1 | - | 5 - 6 | 681 | 71 | |
| AK031533.1 | - | 690 | 71 | |||
| AK087952.1 | - | 5 - 6 | 687 | 71 | ||
| AK132434.1 | - | 5 - 6 | 694 | 71 | ||
| AK145925.1 | - | 5 - 6 | 682 | 71 | ||
| BC021341.1 | - | 5 - 6 | 678 | 5066146 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Xab2
Genomic Map