Assay Details
| Gene Symbol: Xab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026156.2 | NP_080432.1 | 16 - 17 | 2283 | 95 | |
| GenBank mRNA | AK002890.1 | - | 16 - 17 | 2284 | 95 | |
| AK008628.1 | - | 1 - 2 | 185 | 95 | ||
| AK031533.1 | - | 2370 | 95 | |||
| AK087952.1 | - | 13 - 14 | 2546 | 95 | ||
| AK132434.1 | - | 16 - 17 | 2297 | 95 | ||
| AK145925.1 | - | 16 - 17 | 2285 | 95 | ||
| BC021341.1 | - | 16 - 17 | 2281 | 5066146 | 95 | |
More Information
Set Membership: |
|

Xab2
Genomic Map