Assay Details
| Gene Symbol: Plxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026162.6 | NP_080438.2 | 6 - 7 | 1608 | 76 | |
| GenBank mRNA | AF378761.1 | - | 6 - 7 | 807 | 76 | |
| AK004640.1 | - | 6 - 7 | 1590 | 76 | ||
| AK019482.2 | - | 6 - 7 | 1564 | 76 | ||
| AK035165.1 | - | 6 - 7 | 1610 | 76 | ||
| AK154921.1 | - | 6 - 7 | 1555 | 76 | ||
| BC057881.1 | - | 6 - 7 | 868 | 5349869 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plxdc2
Genomic Map