Assay Details
| Gene Symbol: Gpatch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026181.1 | NP_080457.1 | 14 - 15 | 2055 | 100 | |
| GenBank mRNA | AK004868.1 | - | 14 - 15 | 2055 | 100 | |
| AK014367.1 | - | 14 - 15 | 2034 | 100 | ||
| AK154571.1 | - | 14 - 15 | 2036 | 100 | ||
| BC026394.1 | - | 8 - 9 | 1221 | 100 | ||
| BC100577.1 | - | 14 - 15 | 2030 | 100 | ||
| BC125584.1 | - | 14 - 15 | 2046 | 40130099 | 100 | |
| BC132155.1 | - | 14 - 15 | 2046 | 40130432 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpatch1
Genomic Map