Assay Details
| Gene Symbol: Slc39a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135149.1 | NP_001128621.1 | 3 - 4 | 755 | 89 | |
| NM_001135150.1 | NP_001128622.1 | 3 - 4 | 905 | 89 | ||
| NM_026228.5 | NP_080504.3 | 3 - 4 | 864 | 89 | ||
| GenBank mRNA | AK014895.1 | - | 3 - 4 | 868 | 89 | |
| AK016853.1 | - | 3 - 4 | 642 | 89 | ||
| AK028215.1 | - | 3 - 4 | 653 | 89 | ||
| AK085740.1 | - | 3 - 4 | 759 | 89 | ||
| BC006731.1 | - | 2 - 3 | 566 | 3155670 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a8
Genomic Map