Assay Details
| Gene Symbol: Narfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026238.4 | NP_080514.3 | 8 - 9 | 935 | 84 | |
| GenBank mRNA | AK003464.1 | - | 3 - 4 | 268 | 84 | |
| AK014038.1 | - | 8 - 9 | 919 | 84 | ||
| AK018548.1 | - | 8 - 9 | 906 | 84 | ||
| AK043694.1 | - | 8 - 9 | 912 | 84 | ||
| BC020468.1 | - | 8 - 9 | 895 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Narfl
Genomic Map