Assay Details
| Gene Symbol: Nsmce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026330.3 | NP_080606.2 | 3 - 4 | 404 | 84 | |
| GenBank mRNA | AK009715.1 | - | 3 - 4 | 293 | 84 | |
| AK011011.1 | - | 3 - 4 | 309 | 84 | ||
| AK160217.1 | - | 3 - 4 | 304 | 84 | ||
| AK166410.1 | - | 3 - 4 | 297 | 84 | ||
| BC031848.1 | - | 3 - 4 | 383 | 3984224 | 84 | |
| BC049558.1 | - | 3 - 4 | 397 | 6707952 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nsmce1
Genomic Map