Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800019673.mm.1 |
NM_026423.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019673.mm.1 |
NM_026423.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019673.mm.1 |
TC0800002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019669.mm.1 |
NM_026423.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019669.mm.1 |
NM_026423.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019669.mm.1 |
TC0800002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019671.mm.1 |
NM_026423.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019671.mm.1 |
NM_026423.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019671.mm.1 |
TC0800002579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019672.mm.1 |
NM_026423.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019672.mm.1 |
NM_026423.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019672.mm.1 |
TC0800002579.mm.1 |
MTA TC |