| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00000881.mm.1 |
NM_001166635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000881.mm.1 |
NM_001166635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000881.mm.1 |
NM_026524.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000881.mm.1 |
NM_026524.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000881.mm.1 |
TC0X00000131.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000882.mm.1 |
NM_001166635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000882.mm.1 |
NM_001166635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000882.mm.1 |
NM_026524.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000882.mm.1 |
NM_026524.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000882.mm.1 |
TC0X00000131.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000889.mm.1 |
NM_001166635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000889.mm.1 |
NM_001166635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000889.mm.1 |
NM_026524.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000889.mm.1 |
NM_026524.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000889.mm.1 |
TC0X00000131.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000883.mm.1 |
NM_001166635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000883.mm.1 |
NM_001166635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000883.mm.1 |
NM_026524.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000883.mm.1 |
NM_026524.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000883.mm.1 |
TC0X00000131.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000888.mm.1 |
NM_001166635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000888.mm.1 |
NM_001166635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000888.mm.1 |
NM_026524.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000888.mm.1 |
NM_026524.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000888.mm.1 |
TC0X00000131.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00000886.mm.1 |
NM_001166635.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000886.mm.1 |
NM_001166635.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000886.mm.1 |
NM_026524.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000886.mm.1 |
NM_026524.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00000886.mm.1 |
TC0X00000131.mm.1 |
MTA TC |