Assay Details
| Gene Symbol: N6amt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026526.2 | NP_080802.1 | 2 | 71 | 75 | |
| GenBank mRNA | AK005312.1 | - | 2 | 65 | 75 | |
| AK012083.1 | - | 2 | 72 | 75 | ||
| BC051925.1 | - | 2 | 62 | 5691329 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

N6amt2
Genomic Map