Assay Details
| Gene Symbol: Wdr86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081441.1 | NP_001074910.1 | 6 | 1386 | 101 | |
| GenBank mRNA | AK019251.1 | - | 1 | 153 | 101 | |
| AK019267.1 | - | 1 | 153 | 101 | ||
| BC034840.1 | - | 1 | 709 | 101 | ||
| BC048684.1 | - | 2 | 675 | 101 | ||
| BC059069.1 | - | 5 | 3958 | 101 | ||
| BC132276.1 | - | 1 | 147 | 101 | ||
| BC132278.1 | - | 1 | 147 | 101 | ||
| BY712199.1 | - | 1 | 147 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | BC048684.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | BC059069.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | BY712199.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | NM_001081441.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | AK019251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | BC048684.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | BC059069.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | BY712199.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | NM_001081441.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P372456 | AK019251.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602414 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21750 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358761 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56925 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | BC048684.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | BC059069.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | BY712199.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | NM_001081441.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | AK019251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | BC048684.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | BC059069.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | BY712199.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | NM_001081441.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P372456 | AK019251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | BC048684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | BC059069.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | BY712199.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | NM_001081441.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | AK019251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | BC048684.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | BC059069.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | BY712199.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | NM_001081441.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P372456 | AK019251.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23543533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04636340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04682450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04923374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04923375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04874567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04660069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04119419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04119412 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04398624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04804311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04730930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04119410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04892248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04119411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04405174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23543614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04128194 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10528681 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528684 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528683 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528680 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10528685 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419464_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528679 | - | - |
| Mouse Genome 430 2.0 Array | 1419464_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419464_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269633_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 269633_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500019057.mm.1 | NM_001081441.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019057.mm.1 | NM_001081441.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019057.mm.1 | TC0500002091.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019058.mm.1 | NM_001081441.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019058.mm.1 | NM_001081441.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019058.mm.1 | TC0500002091.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419464_PM_at | - | - |
| Mouse Expression Set 430 | 1419464_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419464_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419464_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419464_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419464_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223387 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223387 | - | - |

Wdr86
Genomic Map