Assay Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026660.2 | NP_080936.1 | 8 - 9 | 1120 | 79 | |
| GenBank mRNA | AK003763.1 | - | 3 - 4 | 388 | 79 | |
| AK018736.1 | - | 8 - 9 | 1133 | 79 | ||
| AK089055.1 | - | 8 - 9 | 1137 | 79 | ||
| AK152438.1 | - | 8 - 9 | 1106 | 79 | ||
| BC023012.1 | - | 8 - 9 | 1120 | 5376230 | 79 | |
| BC064775.1 | - | 8 - 9 | 1018 | 30023877 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd10
Genomic Map