Assay Details
| Gene Symbol: Dhrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027790.2 | NP_082066.2 | 10 - 11 | 968 | 76 | |
| GenBank mRNA | AK017274.1 | - | 10 - 11 | 970 | 76 | |
| BC117731.2 | - | 9 - 10 | 861 | 40089799 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962139 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 314485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544910 | AK011491.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908315 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267435 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476094 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86779 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146002 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163306 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29616885 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08117992 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163306 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146002 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02853362 | - | - |
| Whole Mouse Genome G4122A | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174215 | NM_027790.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P174215 | AK017274.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107192 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128996 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146002 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29617019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29616973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11283768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11405836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11420594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11458533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11435673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11495154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11548280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11415742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11415756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11695612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29617015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11405827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11319247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29616948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10415175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415167 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415172 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415166 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415165 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415173 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415168 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415164 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421663_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415163 | - | - |
| Mouse Genome 430 2.0 Array | 1421663_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421663_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71412_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71412_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400006071.mm.1 | NM_027790.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006071.mm.1 | NM_027790.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006071.mm.1 | TC1400000776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400006070.mm.1 | NM_027790.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006070.mm.1 | NM_027790.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006070.mm.1 | TC1400000776.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400006072.mm.1 | NM_027790.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006072.mm.1 | NM_027790.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006072.mm.1 | TC1400000776.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421663_at | - | - |
| Mouse Expression Set 430 | 1421663_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421663_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421663_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421663_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421663_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 390549 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4200577 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004200577 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2639136 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2639136 | - | - |
| mouse-6 v1.1 (Illumina) | scl46285.11.1_214-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4200577 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46285.11.1_214-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4200577 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2639136 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2639136 | - | - |

Dhrs2
Genomic Map