Assay Details
| Gene Symbol: Mus81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027877.3 | NP_082153.3 | 5 - 6 | 737 | 56 | |
| GenBank mRNA | AF425647.1 | - | 5 - 6 | 552 | 56 | |
| AK004647.1 | - | 4 - 5 | 420 | 56 | ||
| AK145858.1 | - | 5 - 6 | 687 | 56 | ||
| AK205475.1 | - | 3 - 4 | 200 | 56 | ||
| BC026560.1 | - | 5 - 6 | 750 | 5038349 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mus81
Genomic Map