| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700005554.mm.1 |
NM_028120.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005554.mm.1 |
NM_028120.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005554.mm.1 |
TC0700000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005548.mm.1 |
NM_028120.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005548.mm.1 |
NM_028120.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005548.mm.1 |
TC0700000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005551.mm.1 |
NM_028120.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005551.mm.1 |
NM_028120.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005551.mm.1 |
TC0700000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005550.mm.1 |
NM_028120.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005550.mm.1 |
NM_028120.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005550.mm.1 |
TC0700000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005547.mm.1 |
NM_028120.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005547.mm.1 |
NM_028120.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005547.mm.1 |
TC0700000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005544.mm.1 |
TC0700000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005549.mm.1 |
NM_028120.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005549.mm.1 |
NM_028120.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005549.mm.1 |
TC0700000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700005552.mm.1 |
NM_028120.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005552.mm.1 |
NM_028120.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700005552.mm.1 |
TC0700000660.mm.1 |
MTA TC |