Assay Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028803.4 | NP_083079.1 | 9 - 10 | 1442 | 75 | |
| GenBank mRNA | AK009815.1 | - | 9 - 10 | 1258 | 75 | |
| AK050365.1 | - | 9 - 10 | 1329 | 75 | ||
| AK050423.1 | - | 9 - 10 | 1328 | 75 | ||
| AK148186.1 | - | 3 - 4 | 370 | 75 | ||
| BC017541.1 | - | 9 - 10 | 1303 | 4526961 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gbe1
Genomic Map