Assay Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028803.4 | NP_083079.1 | 10 - 11 | 1530 | 120 | |
| GenBank mRNA | AK009815.1 | - | 10 - 11 | 1346 | 120 | |
| AK050365.1 | - | 10 - 11 | 1417 | 120 | ||
| AK050423.1 | - | 10 - 11 | 1416 | 120 | ||
| AK148186.1 | - | 4 - 5 | 458 | 120 | ||
| BC017541.1 | - | 10 - 11 | 1391 | 4526961 | 120 | |
More Information
Set Membership: |
|

Gbe1
Genomic Map