Assay Details
| Gene Symbol: Rnf138rt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028842.3 | NP_083118.1 | 1 | 752 | 81 | |
| GenBank mRNA | AK006700.1 | - | 1 | 751 | 81 | |
| BC125311.1 | - | 1 | 680 | 81 | ||
| BC125313.1 | - | 1 | 680 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | NR_003640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | AK006700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | BC125311.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | BC125313.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | NR_003640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | AK006700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | BC125311.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P408703 | BC125313.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853321 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | NR_003640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | AK006700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | BC125311.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | BC125313.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | NR_003640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | AK006700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | BC125311.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P408703 | BC125313.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | NR_003640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | AK006700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | BC125311.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | BC125313.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | NR_003640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | AK006700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | BC125311.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P408703 | BC125313.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15222682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14639781 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794854 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794899 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794865 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794841 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14803834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14803839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15064303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14822429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32794872 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607722 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449239_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607718 | - | - |
| Mouse Genome 430 2.0 Array | 1449239_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449239_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74264_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74264_PM_at | - | - |
| Mouse Expression Set 430 | 1449239_at | - | - |
| Mouse Expression Set 430 | 1449239_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449239_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449239_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449239_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449239_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 841452 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2597397 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2597397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2967255 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2967255 | - | - |

Rnf138rt1
Genomic Map