Assay Details
| Gene Symbol: Prkd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171004.1 | NP_001164475.1 | 4 - 5 | 1321 | 76 | |
| NM_001171005.1 | NP_001164476.1 | 3 - 4 | 517 | 76 | ||
| NM_029239.3 | NP_083515.2 | 4 - 5 | 1321 | 76 | ||
| GenBank mRNA | AK016167.1 | - | 4 - 5 | 476 | 76 | |
| AK030058.1 | - | 3 - 4 | 514 | 76 | ||
| AK050059.1 | - | 2 - 3 | 345 | 76 | ||
| BC027320.1 | - | 3 - 4 | 720 | 76 | ||
| BC037012.1 | - | 3 - 4 | 965 | 5342428 | 76 | |
| BC089164.1 | - | 4 - 5 | 1220 | 5400250 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkd3
Genomic Map