Assay Details
| Gene Symbol: Cxcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030712.4 | NP_109637.3 | 1 - 2 | 72 | 90 | |
| GenBank mRNA | AF301018.1 | - | 1 - 2 | 75 | 90 | |
| AF305709.1 | - | 1 - 2 | 75 | 90 | ||
| AK052901.1 | - | 1 - 2 | 70 | 90 | ||
| AK137703.1 | - | 1 - 2 | 51 | 90 | ||
| AK153914.1 | - | 1 - 2 | 71 | 90 | ||
| JC209414.1 | - | 1 - 2 | 72 | 90 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF301018.1,AF305709.1,AK052901.1,AK137703.1,AK153914.1,JC209414.1,NM_030712.4 transcripts.
Set Membership: |
|

Cxcr6
Genomic Map