Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100036688.mm.1 |
NM_001130187.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036688.mm.1 |
NM_001130187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036688.mm.1 |
NM_031878.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036688.mm.1 |
NM_031878.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036688.mm.1 |
TC1100003918.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036691.mm.1 |
NM_001130187.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036691.mm.1 |
NM_001130187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036691.mm.1 |
NM_031878.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036691.mm.1 |
NM_031878.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036691.mm.1 |
TC1100003918.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036689.mm.1 |
NM_001130187.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036689.mm.1 |
NM_001130187.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036689.mm.1 |
NM_031878.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036689.mm.1 |
NM_031878.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036689.mm.1 |
TC1100003918.mm.1 |
MTA TC |