Assay Details
| Gene Symbol: Arpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177615.1 | NP_001171086.1 | 3 - 4 | 535 | 63 | |
| NM_001177616.1 | NP_001171087.1 | 4 - 5 | 542 | 63 | ||
| NM_001177617.1 | NP_001171088.1 | 4 - 5 | 709 | 63 | ||
| NM_001177618.1 | NP_001171089.1 | 3 - 4 | 240 | 63 | ||
| NM_001177619.1 | NP_001171090.1 | 4 - 5 | 907 | 63 | ||
| NM_001177620.1 | NP_001171091.1 | 5 - 6 | 482 | 63 | ||
| NM_001177623.1 | NP_001171094.1 | 4 - 5 | 471 | 63 | ||
| NM_028755.3 | NP_083031.1 | 4 - 5 | 444 | 63 | ||
| NM_033264.2 | NP_150289.1 | 3 - 4 | 375 | 63 | ||
| GenBank mRNA | AF324451.1 | - | 3 - 4 | 335 | 63 | |
| AK002950.2 | - | 4 - 5 | 428 | 63 | ||
| AK029467.1 | - | 4 - 5 | 908 | 63 | ||
| AK032417.1 | - | 4 - 5 | 468 | 63 | ||
| AK032543.1 | - | 2 - 3 | 155 | 63 | ||
| AK034376.1 | - | 2 - 3 | 88 | 63 | ||
| AK045507.1 | - | 4 - 5 | 909 | 63 | ||
| AK050613.1 | - | 4 - 5 | 472 | 63 | ||
| AK137974.1 | - | 4 - 5 | 710 | 63 | ||
| AK153892.1 | - | 5 - 6 | 1190 | 63 | ||
| AK159004.1 | - | 3 - 4 | 514 | 63 | ||
| AK160860.1 | - | 5 - 6 | 480 | 63 | ||
| BC027107.1 | - | 4 - 5 | 432 | 63 | ||
| BC053001.1 | - | 2 - 3 | 206 | 5686446 | 63 | |
| BC098368.1 | - | 4 - 5 | 489 | 63 | ||
| BC128376.1 | - | 3 - 4 | 256 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK159004.1, AK160860.1 transcripts.
Set Membership: |
|

Arpp21
Genomic Map