Assay Details
| Gene Symbol: Arid1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080819.1 | NP_001074288.1 | 3 - 4 | 1807 | 77 | |
| GenBank mRNA | AF268912.1 | - | 2 - 3 | 658 | 77 | |
| AK142143.1 | - | 3 - 4 | 735 | 77 | ||
| BC082554.1 | - | 1039 | 30622048 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid1a
Genomic Map