Assay Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102471.1 | NP_001095941.1 | 2 - 3 | 1928 | 108 | |
| NM_033569.3 | NP_291047.2 | 2 - 3 | 1928 | 108 | ||
| GenBank mRNA | AF216961.1 | - | 2 - 3 | 1240 | 108 | |
| AK032006.1 | - | 2 - 3 | 1887 | 108 | ||
| AK053711.1 | - | 2 - 3 | 1946 | 108 | ||
| AK159616.1 | - | 2 - 3 | 1934 | 108 | ||
| BC125033.1 | - | 2 - 3 | 1240 | 108 | ||
| GQ225745.1 | - | 2 - 3 | 1227 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm2
Genomic Map