| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400007924.mm.1 |
NM_001080926.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007924.mm.1 |
NM_001080926.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007924.mm.1 |
NR_033496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007924.mm.1 |
NR_033496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007924.mm.1 |
TC0400001135.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400007923.mm.1 |
NM_001080926.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007923.mm.1 |
NM_001080926.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007923.mm.1 |
NR_033496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007923.mm.1 |
NR_033496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007923.mm.1 |
TC0400001135.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400007926.mm.1 |
NM_001080926.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007926.mm.1 |
NM_001080926.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007926.mm.1 |
NR_033496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007926.mm.1 |
NR_033496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007926.mm.1 |
TC0400001135.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400007922.mm.1 |
NM_001080926.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007922.mm.1 |
NM_001080926.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007922.mm.1 |
NR_033496.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007922.mm.1 |
NR_033496.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400007922.mm.1 |
TC0400001135.mm.1 |
MTA TC |