Assay Details
| Gene Symbol: Adgra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054044.2 | NP_473385.2 | 13 - 14 | 2157 | 86 | |
| GenBank mRNA | AF378759.1 | - | 13 - 14 | 2157 | 86 | |
| AK018412.1 | - | 4 - 5 | 569 | 86 | ||
| BC138451.1 | - | 13 - 14 | 2147 | 8861471 | 86 | |
| BC138452.1 | - | 13 - 14 | 2147 | 8861472 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgra3
Genomic Map