Assay Details
| Gene Symbol: Slc8a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167920.1 | NP_001161392.1 | 4 - 5 | 2614 | 80 | |
| NM_080440.3 | NP_536688.2 | 5 - 6 | 2635 | 80 | ||
| GenBank mRNA | AF453257.1 | - | 5 - 6 | 2636 | 80 | |
| AK139260.1 | - | 4 - 5 | 2601 | 80 | ||
| AK165197.1 | - | 4 - 5 | 2400 | 80 | ||
| BC052435.1 | - | 4 - 5 | 2385 | 6837128 | 80 | |
| BC080862.1 | - | 4 - 5 | 2102 | 30357598 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc8a3
Genomic Map