Assay Details
| Gene Symbol: Slc8a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167920.1 | NP_001161392.1 | 5 - 6 | 2710 | 71 | |
| NM_080440.3 | NP_536688.2 | 6 - 7 | 2731 | 71 | ||
| GenBank mRNA | AF453257.1 | - | 6 - 7 | 2732 | 71 | |
| AK139260.1 | - | 5 - 6 | 2697 | 71 | ||
| AK165197.1 | - | 5 - 6 | 2496 | 71 | ||
| BC052435.1 | - | 5 - 6 | 2481 | 6837128 | 71 | |
| BC080862.1 | - | 5 - 6 | 2198 | 30357598 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc8a3
Genomic Map