Assay Details
| Gene Symbol: Bsnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080458.2 | NP_536706.2 | 1 - 2 | 374 | 63 | |
| GenBank mRNA | AF391088.1 | - | 1 - 2 | 371 | 63 | |
| AK052587.1 | - | 1 - 2 | 375 | 63 | ||
| AY373833.1 | - | 1 - 2 | 251 | 63 | ||
| BC038287.1 | - | 1 - 2 | 425 | 4221532 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bsnd
Genomic Map