Assay Details
| Gene Symbol: Smc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080470.1 | NP_536718.1 | 23 - 24 | 3510 | 67 | |
| GenBank mRNA | AF303827.1 | - | 23 - 24 | 3510 | 67 | |
| BC129800.1 | - | 23 - 24 | 3501 | 40048794 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smc1b
Genomic Map