Assay Details
| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287172.1 | NP_001274101.1 | 4 - 5 | 562 | 65 | |
| NM_001287176.1 | NP_001274105.1 | 4 - 5 | 685 | 65 | ||
| NM_007431.3 | NP_031457.2 | 4 - 5 | 537 | 65 | ||
| GenBank mRNA | AB473959.1 | - | 3 - 4 | 301 | 65 | |
| AK076459.1 | - | 4 - 5 | 472 | 65 | ||
| AK161780.1 | - | 4 - 5 | 473 | 65 | ||
| AK164002.1 | - | 4 - 5 | 530 | 65 | ||
| AK166732.1 | - | 4 - 5 | 471 | 65 | ||
| AK167023.1 | - | 4 - 5 | 472 | 65 | ||
| AK167481.1 | - | 4 - 5 | 472 | 65 | ||
| BC065175.1 | - | 4 - 5 | 460 | 6807509 | 65 | |
| CN539670.1 | - | 4 - 5 | 564 | 65 | ||
| J02980.1 | - | 4 - 5 | 476 | 65 | ||
| X13409.1 | - | 4 - 5 | 420 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alpl
Genomic Map