Assay Details
| Gene Symbol: Ap1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007457.2 | NP_031483.1 | 3 - 4 | 339 | 102 | |
| GenBank mRNA | AK002225.1 | - | 3 - 4 | 317 | 102 | |
| BC052692.1 | - | 3 - 4 | 343 | 5714625 | 102 | |
| M62418.1 | - | 3 - 4 | 339 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993268 | M62418.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1993268 | NM_007457.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992257 | M62418.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992257 | NM_007457.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992257 | AK002225.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1992257 | BC052692.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202256 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32263 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146564 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102568 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993268 | M62418.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1993268 | NM_007457.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992257 | M62418.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992257 | NM_007457.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992257 | AK002225.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1992257 | BC052692.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124301 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24124309 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102568 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05759683 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01231228 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146564 | - | - |
| Whole Mouse Genome G4122A | A_51_P166394 | M62418.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P166394 | NM_007457.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P166394 | AK002225.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P166394 | BC052692.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139399 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102568 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146564 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04729371 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04729376 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04729384 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04729372 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04425642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24124201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04151785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04425641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04729373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04729385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04729382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04214925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124296 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04243445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124326 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04149513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04910293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04214919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04243459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04532470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04560182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04492829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24124299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04149514 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534663 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534661 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534666 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416087_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534660 | - | - |
| Mouse Genome 430 2.0 Array | 1416087_at | - | - |
| Mouse Genome 430 2.0 Array | 1416087_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11769_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11769_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031079.mm.1 | NM_007457.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031079.mm.1 | NM_007457.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031079.mm.1 | TC0500003430.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416087_PM_at | - | - |
| Mouse Expression Set 430 | 1416087_at | - | - |
| Murine Genome U74v2 Set | 160552_PM_at | - | - |
| Murine Genome U74v2 Set | 160552_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416087_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416087_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416087_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416087_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 719517 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4570520 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2030471 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6900603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030471 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2634108 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215903 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2748777 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2748777 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215903 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2634108 | - | - |
| mouse-6 v1.1 (Illumina) | scl25892.5.96_2-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0004007.1_23-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003948.1_98-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900603 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570520 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030471 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004007.1_23-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25892.5.96_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003948.1_98-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900603 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570520 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030471 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2634108 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2748777 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1215903 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2634108 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2748777 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215903 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0004007.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25892.5.96_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003948.1_98-S | - | - |

Ap1s1
Genomic Map