Assay Details
| Gene Symbol: Arnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007488.3 | NP_031514.3 | 2 - 3 | 304 | 60 | |
| GenBank mRNA | AB093225.1 | - | 4 - 5 | 484 | 60 | |
| AK032628.1 | - | 2 - 3 | 285 | 60 | ||
| BC054546.1 | - | 2 - 3 | 225 | 6409636 | 60 | |
| BU056629.1 | - | 2 - 3 | 442 | 60 | ||
| D63644.1 | - | 2 - 3 | 180 | 60 | ||
More Information
Set Membership: |
|

Arnt2
Genomic Map