Assay Details
| Gene Symbol: Arnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007488.3 | NP_031514.3 | 10 - 11 | 1245 | 79 | |
| GenBank mRNA | AB093225.1 | - | 12 - 13 | 1425 | 79 | |
| AK032628.1 | - | 10 - 11 | 1226 | 79 | ||
| BC054546.1 | - | 10 - 11 | 1166 | 6409636 | 79 | |
| D63644.1 | - | 10 - 11 | 1121 | 79 | ||
More Information
Set Membership: |
|

Arnt2
Genomic Map