Assay Details
| Gene Symbol: Atf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007498.3 | NP_031524.2 | 2 - 3 | 489 | 61 | |
| GenBank mRNA | AB291912.1 | - | 418 | 61 | ||
| AK133965.1 | - | 2 - 3 | 493 | 61 | ||
| AK156786.1 | - | 2 - 3 | 409 | 61 | ||
| AY329367.1 | - | 2 - 3 | 158 | 61 | ||
| BC019946.1 | - | 2 - 3 | 500 | 4011514 | 61 | |
| BC064799.1 | - | 2 - 3 | 355 | 30054998 | 61 | |
| CT010291.1 | - | 1 - 2 | 244 | 61 | ||
| D50524.1 | - | 1 - 2 | 244 | 61 | ||
| U19118.1 | - | 2 - 3 | 411 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Transcriptional response to corticotropin-releasing factor in AtT-20 cells.
- Authors
- Peeters PJ, Gohlmann HW, Van den Wyngaert I, Swagemakers SM, Bijnens L, Kass SU, Steckler T
- Journal
- Mol Pharmacol - Page No 1083 (Mon Nov 01 00:00:00 UTC 2004)
- Volume (Year)
- 66 (2004)
- Product Usage
- -

Atf3
Genomic Map