Assay Details
| Gene Symbol: Trmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080999.2 | NP_001074468.1 | 3 - 4 | 1030 | 103 | |
| NM_001081000.2 | NP_001074469.1 | 3 - 4 | 1077 | 103 | ||
| NM_001195205.1 | NP_001182134.1 | 3 - 4 | 1030 | 103 | ||
| GenBank mRNA | AK034102.1 | - | 3 - 4 | 910 | 103 | |
| AK080170.1 | - | 3 - 4 | 905 | 103 | ||
| AK133844.1 | - | 3 - 4 | 905 | 103 | ||
| AK134368.1 | - | 3 - 4 | 1031 | 103 | ||
| AK146391.1 | - | 3 - 4 | 970 | 103 | ||
| AK150482.1 | - | 3 - 4 | 875 | 103 | ||
| AK151311.1 | - | 3 - 4 | 875 | 103 | ||
| AK151422.1 | - | 3 - 4 | 901 | 103 | ||
| AK151725.1 | - | 3 - 4 | 875 | 103 | ||
| AK151778.1 | - | 3 - 4 | 875 | 103 | ||
| AK152055.1 | - | 3 - 4 | 875 | 103 | ||
| AK152201.1 | - | 3 - 4 | 875 | 103 | ||
| AK152363.1 | - | 3 - 4 | 875 | 103 | ||
| AK152548.1 | - | 3 - 4 | 875 | 103 | ||
| AK152903.1 | - | 3 - 4 | 875 | 103 | ||
| AK153043.1 | - | 3 - 4 | 875 | 103 | ||
| AK153190.1 | - | 3 - 4 | 901 | 103 | ||
| AK169240.1 | - | 3 - 4 | 895 | 103 | ||
| AK186624.1 | - | 2 - 3 | 247 | 103 | ||
| BC027431.1 | - | 2 - 3 | 1142 | 3156359 | 103 | |
| BC038486.1 | - | 3 - 4 | 903 | 3155739 | 103 | |
| BC046976.1 | - | 3 - 4 | 911 | 6337243 | 103 | |
| X56044.1 | - | 2 - 3 | 1474 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trmt2a
Genomic Map