Assay Details
| Gene Symbol: Trmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080999.2 | NP_001074468.1 | 9 - 10 | 1749 | 62 | |
| NM_001081000.2 | NP_001074469.1 | 9 - 10 | 1796 | 62 | ||
| GenBank mRNA | AK034102.1 | - | 6 - 7 | 1945 | 62 | |
| AK133844.1 | - | 9 - 10 | 1624 | 62 | ||
| AK134368.1 | - | 9 - 10 | 1750 | 62 | ||
| AK146391.1 | - | 9 - 10 | 1689 | 62 | ||
| AK150482.1 | - | 9 - 10 | 1594 | 62 | ||
| AK151311.1 | - | 9 - 10 | 1594 | 62 | ||
| AK151422.1 | - | 9 - 10 | 1620 | 62 | ||
| AK151725.1 | - | 9 - 10 | 1594 | 62 | ||
| AK151778.1 | - | 9 - 10 | 1594 | 62 | ||
| AK152055.1 | - | 9 - 10 | 1594 | 62 | ||
| AK152201.1 | - | 9 - 10 | 1594 | 62 | ||
| AK152363.1 | - | 9 - 10 | 1594 | 62 | ||
| AK152548.1 | - | 9 - 10 | 1594 | 62 | ||
| AK152903.1 | - | 9 - 10 | 1594 | 62 | ||
| AK153043.1 | - | 9 - 10 | 1594 | 62 | ||
| AK153190.1 | - | 9 - 10 | 1620 | 62 | ||
| BC027431.1 | - | 8 - 9 | 1861 | 3156359 | 62 | |
| BC038486.1 | - | 9 - 10 | 1622 | 3155739 | 62 | |
| X56044.1 | - | 8 - 9 | 2193 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trmt2a
Genomic Map