Assay Details
| Gene Symbol: Nipsnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008698.2 | NP_032724.1 | 6 - 7 | 587 | 78 | |
| GenBank mRNA | AJ001260.1 | - | 6 - 7 | 587 | 78 | |
| AK013140.1 | - | 6 - 7 | 595 | 78 | ||
| AK031804.1 | - | 6 - 7 | 603 | 78 | ||
| BC010837.1 | - | 6 - 7 | 585 | 4242309 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nipsnap1
Genomic Map